Merck
[Pathogenetic bases of dietary prevention of dichlorobutene poisoning].
Voprosy pitaniia (1987-09-01)
P A Bakalian, O A Antonian
ABSTRACT
It was shown experimentally that alimentary prevention of dichlorobutene intoxication could be realized in the presence of a protein-enriched ration, through additional injections of nutrients possessing antioxidant and lipotropic properties and playing a pathogenetically valid protective role in this intoxication.