Merck
  • Kinetic and analytical study of the photo-induced degradation of monuron by nitrates and nitrites under irradiation or in the dark.
Kinetic and analytical study of the photo-induced degradation of monuron by nitrates and nitrites under irradiation or in the dark.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology (2012-06-05)
Hadjira Boucheloukh, Tahar Sehili, Nadia Kouachi, Kamel Djebbar
ABSTRACT
The photo-induced transformation of monuron (3-(4-chlorophenyl)-1,1 dimethylurea) was investigated in an aqueous solution containing nitrates and nitrites at 310 nm and 365 nm, respectively. In both NO(3)(-) and NO(2)(-) conditions, the degradation of monuron followed pseudo-first order kinetics. The intermediate products were identified by GC-MS, and the nitration, hydroxylation and coupling reactions were determined. In addition, the oxidation of the N-terminus group, the substitution of chlorine by ˙OH and the nitration by ˙NO(2) radical onto the phenyl ring were observed. The photo-induced transformation of monuron was studied under variable conditions of pH, inducer concentration, substrate concentration, humic acids, oxygen content and salts used as hydroxyl radical scavengers. The photodegradation rates were strongly influenced by all the above parameters. The degradation of monuron was also studied in the dark and in the presence of NO(2)(-) as well as in an aqueous solution with the addition of hydrogen peroxide.