Merck
  • Aligned carbon nanotubes sandwiched in epitaxial NbC film for enhanced superconductivity.
Aligned carbon nanotubes sandwiched in epitaxial NbC film for enhanced superconductivity.
Nanoscale (2012-03-01)
Yingying Zhang, Filip Ronning, Krzysztof Gofryk, Nathan A Mara, Nestor Haberkorn, Guifu Zou, Haiyan Wang, Joon H Lee, Eve Bauer, Thomas M McCleskey, Anthony K Burell, Leonardo Civale, Y T Zhu, Quanxi Jia
ABSTRACT
Highly aligned carbon nanotube (CNT) ribbons were sandwiched in epitaxial superconducting NbC films by a chemical solution deposition method. The incorporation of aligned long CNTs into NbC film enhances the normal-state conductivity and improves the superconducting properties of the assembly.