Merck
  • An efficient and selective way to new highly functionalized coronands or spiro derivatives using ultrasonic irradiation.
An efficient and selective way to new highly functionalized coronands or spiro derivatives using ultrasonic irradiation.
Ultrasonics sonochemistry (2011-09-13)
Gheorghita Zbancioc, Ondina Florea, Peter G Jones, Ionel I Mangalagiu
ABSTRACT
A new, selective, straightforward and general method for preparation of highly functionalized coronands or spiro derivatives bearing 1,2-dihydroxyacetophenone unit, under conventional conditions and ultrasonic irradiation, is reported. The reaction setup involves only one step, acylation of an α-chloro-3,4-dihydroxyacetophenone with phthaloyl dichloride derivatives. 1,3- and 1,4-Phthaloyl dichloride derivatives leads to coronands only, while 1,2-phthaloyl dichlorides lead either to coronands or to spiro derivatives. A feasible explanation for the different behavior between conventional and ultrasound methods could be the different reaction mechanism involved in the two procedures: tetrahedral nucleophilic substitution under conventional conditions and radical substitution under ultrasound. Ultrasound induces a remarkable acceleration of the reactions (from days to minutes) and, most significantly, the yields are twice as high. A feasible explanation for the efficiency of the reactions under ultrasonic irradiation is presented.
Sigma-Aldrich
2′,6′-Dihydroxyacetophenone, 97%
Supelco
2′,6′-Dihydroxyacetophenone, matrix substance for MALDI-MS, ≥99.5% (HPLC), Ultra pure
Supelco
2′,6′-Dihydroxyacetophenone, matrix substance for MALDI-MS, ≥99.5%