Merck
  • A peroxisomal carrier delivers NAD⁺ and contributes to optimal fatty acid degradation during storage oil mobilization.
A peroxisomal carrier delivers NAD⁺ and contributes to optimal fatty acid degradation during storage oil mobilization.
The Plant journal : for cell and molecular biology (2011-09-08)
Kristin Bernhardt, Sabrina Wilkinson, Andreas P M Weber, Nicole Linka
ABSTRACT
The existence of a transport protein that imports cytosolic NAD(+) into peroxisomes has been controversially discussed for decades. Nevertheless, the biosynthesis of NAD(+) in the cytosol necessitates the import of NAD(+) into peroxisomes for numerous reduction/oxidation (redox) reactions. However, a gene encoding such a transport system has not yet been identified in any eukaryotic organism. Here, we describe the peroxisomal NAD(+) carrier in Arabidopsis. Our candidate gene At2g39970 encodes for a member of the mitochondrial carrier family. We confirmed its peroxisomal localization using fluorescence microscopy. For a long time At2g39970 was assumed to represent the peroxisomal ATP transporter. In this study, we could show that the recombinant protein mediated the transport of NAD(+) . Hence, At2g39970 was named PXN for peroxisomal NAD(+) carrier. The loss of PXN in Arabidopsis causes defects in NAD(+) -dependent β-oxidation during seedling establishment. The breakdown of fatty acid released from storage oil was delayed, which led to the retention of oil bodies in pxn mutant seedlings. Based on our results, we propose that PXN delivers NAD(+) for optimal fatty acid degradation during storage oil mobilization.