Merck
  • Metabolism studies of the Kratom alkaloids mitraciliatine and isopaynantheine, diastereomers of the main alkaloids mitragynine and paynantheine, in rat and human urine using liquid chromatography-linear ion trap-mass spectrometry.
Metabolism studies of the Kratom alkaloids mitraciliatine and isopaynantheine, diastereomers of the main alkaloids mitragynine and paynantheine, in rat and human urine using liquid chromatography-linear ion trap-mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2011-04-01)
Anika A Philipp, Dirk K Wissenbach, Armin A Weber, Josef Zapp, Hans H Maurer
ABSTRACT
Mitragyna speciosa (Kratom in Thai), native in Southeast Asia, is increasingly misused as a herbal drug of abuse. During metabolism studies on the Kratom alkaloids mitragynine, its diastereomers speciogynine and speciociliatine as well as paynantheine in rats and humans, further isomeric compounds were detected in Kratom users' urine. The question arose whether these compounds were formed from the low abundant, isomeric alkaloids mitraciliatine (MC) and isopaynantheine (ISO-PAY). Therefore, the aim of the presented study was to identify using liquid chromatography-linear ion trap-mass spectrometry their phase I and II metabolites in rat urine after administration of pure MC or ISO-PAY, to confirm their formation in humans, and finally to confirm whether the above-mentioned isomeric compounds in human urine represent MC and ISO-PAY and/or their metabolites. The metabolic pathways of both alkaloids in rats were found to be comparable to those of their corresponding diastereomers. In the human urines tested, not all metabolites found in rats could be detected because of the much lower amounts of MC and ISO-PAY in Kratom. However, all the above-mentioned so far unknown isomeric compounds could be identified in the human urine samples as MC, ISO-PAY and/or their metabolites. The used LC separation was also suitable for the differentiation of all other Kratom alkaloids and their metabolites in human urine.