Merck
Structure of (2,3,7,8,12,13,17,18-octaethylporphinato)copper(II).
Acta crystallographica. Section C, Crystal structure communications (1991-02-15)
R Pak, W R Scheidt
ABSTRACT
[Cu(C36H44N4)], Mr = 596.3, triclinic, P1, a = 13.314(5), b = 13.392(5), c = 4.805(3) A, alpha = 92.42(4), beta = 93.38(4), gamma = 113.08(1) degrees, V = 784.8 A3, Z = 1, Dx = 1.26 g cm-3, Dm = 1.25 g cm-3, lambda (Mo K alpha) = 0.71073 A, mu = 7.25 cm-1, F(000) = 317, T = 293 K, R = 0.055 for 2166 unique observed reflections.
Sigma-Aldrich
2,3,7,8,12,13,17,18-Octaethyl-21H,23H-porphine copper(II), Dye content ~95 %