Merck
  • Ion-pair extraction of histamine from biological fluids and tissues for its determination by high-performance liquid chromatography with fluorescence detection.
Ion-pair extraction of histamine from biological fluids and tissues for its determination by high-performance liquid chromatography with fluorescence detection.
Journal of chromatography (1993-04-21)
D Tsikas, R Velásquez, C Toledano, G Brunner
ABSTRACT
A procedure for the selective ion-pair extraction of histamine from various biological fluids and tissues by using the ion-pairing agent bis(2-ethylhexyl)hydrogen-phosphoric acid has been developed. This procedure enabled routine determination of histamine in plasma, urine and tissue samples of humans and rats by reversed-phase high-performance liquid chromatography with fluorescence detection using o-phthaldialdehyde.
Sigma-Aldrich
Histamine dihydrochloride, ≥99.0% (AT)
Pricing and availability is not currently available.