- Synthesis of new 2-phosphono-alpha-D-glycoside derivatives by stereoselective oxa-Michael addition to a D-galacto derived enone.
Synthesis of new 2-phosphono-alpha-D-glycoside derivatives by stereoselective oxa-Michael addition to a D-galacto derived enone.
Carbohydrate research (2008-04-04)
Francesca Leonelli, Marinella Capuzzi, Enrico Bodo, Pietro Passacantilli, Giovanni Piancatelli
PMID18384762
ABSTRACT
The synthesis of new 2-phosphono-alpha-D-glycoside derivatives by stereoselective oxa-Michael addition to an enone derived from D-galactal and containing a phosphonate group is described. Retro-Michael reactions were prevented by tandem acetylation to trap the unstable enolic intermediates. The stereochemistry of the addition products was established by NOESY experiments and explained with molecular mechanics (MM) and density functional theory (DFT) calculations.