Merck
  • Borrelia burgdorferi bb0728 encodes a coenzyme A disulphide reductase whose function suggests a role in intracellular redox and the oxidative stress response.
Borrelia burgdorferi bb0728 encodes a coenzyme A disulphide reductase whose function suggests a role in intracellular redox and the oxidative stress response.
Molecular microbiology (2006-01-05)
Julie A Boylan, Charles S Hummel, Stéphane Benoit, Jorge Garcia-Lara, Jennifer Treglown-Downey, Edward J Crane, Frank C Gherardini
ABSTRACT
The cellular responses of Borrelia burgdorferiTo reactive oxygen species (ROS) encountered during the different stages of its infective cycle are poorly understood. Few enzymes responsible for protecting proteins, DNA/RNA and lipids from damage by ROS have been identified and characterized. Data presented here suggest that bb0728 encodes an enzyme involved in this process. Biochemical analyses on purified recombinant BB0728 indicated that it functioned as a coenzyme A disulphide reductase (CoADR) (specific activity approximately 26 units per mg of protein). This enzyme was specific for coenzyme A (CoA) disulphide, required NADH and had no significant activity against other disulphides, such as oxidized glutathione or thioredoxin. The high intracellular concentration of reduced CoA (CoASH) in B. burgdorferi cells ( approximately 1 mM) and absence of glutathione suggest that CoA is the major low-molecular-weight thiol in this spirochete. Interestingly, CoASH was able to reduce H(2)O(2) and be regenerated by CoADR suggesting one role for the system may be to protect B. burgdorferi from ROS. Further, mobility-shift assays and transcriptional fusion data indicated that bb0728 was positively regulated by the Borrelia oxidative stress response regulator, BosR. Taken together, these data suggest a role for BB0728 in intracellular redox and the oxidative stress response in B. burgdorferi.
Sigma-Aldrich
Coenzyme A, oxidized lithium salt, ≥85%