Merck
  • Differential accumulation of low-chlorinated (Delor 103) and high-chlorinated (Delor 106) biphenyls in human placental tissue and opposite effects on conversion of DHEA to E2.
Differential accumulation of low-chlorinated (Delor 103) and high-chlorinated (Delor 106) biphenyls in human placental tissue and opposite effects on conversion of DHEA to E2.
Chemosphere (2005-08-27)
R Grabic, L G Hansen, A Ptak, S Crhova, E Ł Gregoraszczuk
ABSTRACT
Polychlorinated biphenyls (PCBs) and related compounds elicit a diverse spectrum of toxic responses. Additionally, they are able to pass through the human placenta. The aim of the presented data was to compare the action of low-chlorinated (Delor 103) and (Delor 106) high-chlorinated biphenyls on placental steroidogenesis. Explants of human placental tissue were used to test differences in PCBs accumulation and influence on placental steroidogenesis. Delor 103 or 106, were added daily for six days at a dose of 200 pg from day 0 to day 6 of culture. The media in the control and experimental groups were changed every day, and collected and frozen for steroid analysis by RIA. Determinations of PCBs of tissue and medium were analysed by GC/MS/MS. Delor 103 was found at a higher level in the tissue than Delor 106. The first day of exposure to Delor 103 had no effect on the conversion of dehydroepiandrosterone (DHEA) to estradiol (E2) while there was a 2-fold decrease in E2 secretion from days 3 to 6. Conversely, Delor 106 caused an immediate increase in E2 secretion, which was maintained at higher levels throughout the exposure period. Differences between the accumulation of lower chlorinated and higher chlorinated biphenyls in human placental tissue and in the properties of the congeners can have multiple effects that may intensify or counteract the effects on uterine contraction by PCBs.
Product Description
Supelco
PCB No 31, analytical standard