Merck
  • Synthesis and biological evaluation of some 5-nitro- and 5-amino derivatives of 2'-deoxycytidine, 2',3'-dideoxyuridine, and 2',3'-dideoxycytidine.
Synthesis and biological evaluation of some 5-nitro- and 5-amino derivatives of 2'-deoxycytidine, 2',3'-dideoxyuridine, and 2',3'-dideoxycytidine.
Nucleosides, nucleotides & nucleic acids (2003-12-19)
Evelina Colacino, Giovanni Sindona, Gilles Gosselin, Christophe Mathé
ABSTRACT
In this article, we describe the synthesis of 5-nitro-1-(2-deoxy-alpha-D-erythro-pentofuranosyl)cytosine (4alpha), 5-nitro-1-(2-deoxy-beta-D-erythro-pentofuranosyl)cytosine (4beta), 5-amino-1-(2-deoxy-alpha-D-erythro-pentofuranosyl)cytosine (5alpha), 5-nitro-1-(2-deoxy-beta-D-erythro-pentofuranosyl)cytosine (5beta), 5-nitro-1-(2,3-dideoxy-beta-D-ribofuranosyl)uracil (6beta), 5-amino-1-(2,3-dideoxy-alpha,beta-D-ribofuranosyl)uracil (7), 5-nitro-1-(2,3-dideoxy-alpha,beta-D-ribofuranosyl)cytosine (8) and 5-amino-1-(2,3-dideoxy-beta-D-ribofuranosyl)cytosine (9beta). The prepared compounds were tested for their activity against HIV and HBV viruses, but they did not show significant activity.