Merck
  • Sex pheromone biosynthesis in the female olive fruit-fly. Double labelling from [18O2]-dioxygen into 1,7-dioxaspiro[5.5]undecane.
Sex pheromone biosynthesis in the female olive fruit-fly. Double labelling from [18O2]-dioxygen into 1,7-dioxaspiro[5.5]undecane.
Chemical communications (Cambridge, England) (2002-07-12)
Mary T Fletcher, Basilis E Mazomenos, John H Georgakopoulos, Maria A Konstantopoulou, Barry J Wood, James J De Voss, William Kitching
ABSTRACT
The demonstration that both oxygen atoms of 1,7-dioxaspiro[5.5]undecane (1), the sex-pheromone of the female olive fly, originate from dioxygen, strongly implicates monooxygenase mediated processes in assembly of (1), and reveals unexpected complexity in the formation of its nine-carbon precursor.
Sigma-Aldrich
1,7-Dioxaspiro[5.5]undecane, ≥97%