- [Method-performance studies of notified analytical method for clofentezine].
[Method-performance studies of notified analytical method for clofentezine].
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan (2002-01-31)
K Sasaki, T Tatsuno, M Nakamura, T Imazawa, O Utsunomiya, Y Kondo, H Osada, K Takahata, M Toyoda
PMID11817146
ABSTRACT
Method-performance studies were conducted for the notified revised analytical method of clofentezine. Clofentezine spiked in azuki beans, apple, orange, banana, grape, tea powder and tea extract at the level of 0.2 microgram/g (2 micrograms/g for tea) was analyzed in replicate in 6 laboratories. Mean values of recovery from 7 crops ranged from 78.4 to 85.2%. Repeatability relative standard deviation values ranged from 2.2 to 4.6% and reproducibility standard deviation values ranged from 4.8 to 10.3%. The detection limits were 0.005-0.01 microgram/g. These results show the notified analytical method has good performance.