Merck
  • A pharmacokinetic/pharmacodynamic approach to show that not all fluoroquinolones exhibit similar sensitivity toward the proconvulsant effect of biphenyl acetic acid in rats.
A pharmacokinetic/pharmacodynamic approach to show that not all fluoroquinolones exhibit similar sensitivity toward the proconvulsant effect of biphenyl acetic acid in rats.
The Journal of antimicrobial chemotherapy (2001-12-26)
S Marchand, C Pariat, A Boulanger, S Bouquet, W Couet
ABSTRACT
The proconvulsant effect of biphenyl acetic acid (BPAA) on several fluoroquinolones (FQs) was investigated in vivo, by measuring drug concentrations in the biophase at the onset of convulsions. Male Sprague-Dawley rats (n = 134) were given BPAA orally, at various doses 1 h before starting FQ infusion, which was maintained until the onset of maximal seizures, when cerebrospinal fluid (CSF) and plasma samples were collected for drug concentration determination. The FQ-BPAA interactions in the biophase (CSF) were adequately described on most occasions by an inhibitory Emax effect model with a baseline effect parameter. The efficacy of the proconvulsant effect was characterized by the ratio of the CSF concentrations of FQs at the onset of convulsant activity when BPAA was absent (CCSF0, FQs) and as BPAA CSF concentrations tended toward infinity (CCSFbase, FQs). This ratio varied from 15 for enoxacin to 1.9 for sparfloxacin. The potency of the proconvulsant effect was characterized by the CSF concentration of BPAA corresponding to a proconvulsant effect half of its maximum. This parameter varied between 0.18 +/- 0.06 micromol/L with enoxacin and 15.0 +/- 12.1 micromol/L with sparfloxacin. The CSF diffusion of all FQs was apparently non-linear, as well as the plasma protein binding of BPAA, complicating interpretation of plasma data. The important variability in the proconvulsant effect of BPAA demonstrated in this study between various FQs suggests that in vitro gamma-aminobutyric acid (GABA) binding experiments conducted in the presence of BPAA are unlikely to be good predictors of FQ convulsant risk in clinical practice.
Sigma-Aldrich
4-Biphenylacetic acid, 98%