Merck
Inhibition of p38 MAP kinase as a therapeutic strategy.
Immunopharmacology (2000-07-06)
J C Lee, S Kumar, D E Griswold, D C Underwood, B J Votta, J L Adams
ABSTRACT
Since the discovery of p38 MAP kinase in 1994, our understanding of its biology has progressed dramatically. The key advances include (1) identification of p38 MAP kinase homologs and protein kinases that act upstream and downstream from p38 MAP kinase, (2) identification of interesting and potentially important substrates, (3) elucidation of the role of p38 MAP kinase in cellular processes and (4) the establishment of the mechanism by which the pyridinylimidazole p38 MAP kinase inhibitors inhibit enzyme activity. It is now known that there are four members of the p38 MAP kinase family. They differ in their tissue distribution, regulation of kinase activation and subsequent phosphorylation of downstream substrates. They also differ in terms of their sensitivities toward the p38 MAP kinase inhibitors. The best-studied isoform is p38 alpha, whose activation has been observed in many hematopoietic and non-hematopoietic cell types upon treatment with appropriate stimuli. The pyridinylimidazole compounds, exemplified by SB 203580, were originally prepared as inflammatory cytokine synthesis inhibitors that subsequently were found to be selective inhibitors of p38 MAP kinase. SB 203580 inhibits the catalytic activity of p38 MAP kinase by competitive binding in the ATP pocket. X-ray crystallographic studies of the target enzyme complexed with inhibitor reinforce the observations made from site-directed mutagenesis studies, thereby providing a molecular basis for understanding the kinase selectivity of these inhibitors. The p38 MAP kinase inhibitors are efficacious in several disease models, including inflammation, arthritis and other joint diseases, septic shock, and myocardial injury. In all cases, p38 activation in key cell types correlated with disease initiation and progression. Treatment with p38 MAP kinase inhibitors attenuated both p38 activation and disease severity. Structurally diverse p38 MAP kinase inhibitors have been tested extensively in preclinical studies.
Sigma-Aldrich
SKF-86002, ≥98% (HPLC), solid
Pricing and availability is not currently available.