- Diverse oxygenations catalyzed by carbazole 1,9a-dioxygenase from Pseudomonas sp. Strain CA10.
Diverse oxygenations catalyzed by carbazole 1,9a-dioxygenase from Pseudomonas sp. Strain CA10.
Carbazole 1,9a-dioxygenase (CARDO) from Pseudomonas sp. strain CA10 is a multicomponent enzyme that catalyzes the angular dioxygenation of carbazole, dibenzofuran, and dibenzo-p-dioxin. It was revealed by gas chromatography-mass spectrometry and 1H and 13C nuclear magnetic resonance analyses that xanthene and phenoxathiin were converted to 2,2',3-trihydroxydiphenylmethane and 2,2',3-trihydroxydiphenyl sulfide, respectively. Thus, for xanthene and phenoxathiin, angular dioxygenation by CARDO occurred at the angular position adjacent to the oxygen atom to yield hetero ring-cleaved compounds. In addition to the angular dioxygenation, CARDO catalyzed the cis dihydroxylation of polycyclic aromatic hydrocarbons and biphenyl. Naphthalene and biphenyl were converted by CARDO to cis-1, 2-dihydroxy-1,2-dihydronaphthalene and cis-2,3-dihydroxy-2, 3-dihydrobiphenyl, respectively. On the other hand, CARDO also catalyzed the monooxygenation of sulfur heteroatoms in dibenzothiophene and of the benzylic methylenic group in fluorene to yield dibenzothiophene-5-oxide and 9-hydroxyfluorene, respectively. These results indicate that CARDO has a broad substrate range and can catalyze diverse oxygenation: angular dioxygenation, cis dihydroxylation, and monooxygenation. The diverse oxygenation catalyzed by CARDO for several aromatic compounds might reflect the differences in the binding of the substrates to the reaction center of CARDO.