Merck
  • Distribution and persistence of tricyaclazole in agricultural field soils.
Distribution and persistence of tricyaclazole in agricultural field soils.
Bulletin of environmental contamination and toxicology (2012-09-28)
Seul Ah Jeong, Shree Prasad Thapa, Hong Ryeol Park, Nam Geon Choi, Jang Hyun Hur
ABSTRACT
Soil is the major sink for majority of pesticides applied on agricultural crops and its fate depends on variety of factors. There is little research on fate of pesticide in field soil under different climatic conditions and there is a need of study on the influence of climate on pesticide degradation and persistence in soil. In the present study, the persistence and distribution of tricyclazole was investigated in rice field soil under the influence of cold winter condition. Field experiment was carried at 35 different field sites from 6 provinces in Republic of Korea. Limit of detection and limit of quantification of tricyclazole were found to be 0.005 and 0.0165 mg/kg, respectively. The concentrations of tricyclazole in soil samples ranged from 0.387 mg/kg in sites in Gyeongsangbuk-do areas and lowest 0.021 mg/kg in sites from Chuncheongnam-do areas. In natural environmental conditions, tricyclazole persisted longer than 11 months post application in agricultural field soils. Our result indicates the influence of cold climatic condition on the persistence of tricyclazole.