Merck
  • Scandium(III)-catalyzed enantioselective allylation of isatins using allylsilanes.
Scandium(III)-catalyzed enantioselective allylation of isatins using allylsilanes.
Organic letters (2012-04-18)
Nadine V Hanhan, Yng C Tang, Ngon T Tran, Annaliese K Franz
ABSTRACT
The scandium(III)-catalyzed enantioselective Hosomi-Sakurai allylation of isatins with various substituted allylic silanes is described. A catalyst loading as low as 0.05 mol % is utilized at room temperature to afford the 3-allyl-3-hydroxy-2-oxindoles in excellent yields and enantioselectivity up to 99% ee, including a demonstration of a gram-scale reaction. The effects of additives and varying silyl groups were explored to demonstrate the scope and application.
Sigma-Aldrich
Scandium, powder, 99.9% trace rare earth metals basis