- Concise formal synthesis of (-)-7-deoxyloganin via N-heterocyclic carbene catalysed rearrangement of α,β-unsaturated enol esters.
Concise formal synthesis of (-)-7-deoxyloganin via N-heterocyclic carbene catalysed rearrangement of α,β-unsaturated enol esters.
Organic & biomolecular chemistry (2011-10-26)
Lisa Candish, David W Lupton
PMID22024770
ABSTRACT
NHC catalysed rearrangement of α,β-unsaturated enol esters derived from formyl acetates and cyclopentyl annulated α,β-unsaturated acids provides the cyclopentapyranone core of (-)-7-deoxyloganin (1) with diastereo- and chemoselectivity in 6 steps starting from (-)-citronellal. The elaboration to the natural product has been investigated using two new approaches. The most successful intercepts our previous work on (-)-7-deoxyloganin (1) allowing completion of a formal total synthesis in 10-steps.
MATERIALS