Merck
  • A possible model of hemoprotein-fatty acid peroxide complex demonstrated by electron spin resonance.
A possible model of hemoprotein-fatty acid peroxide complex demonstrated by electron spin resonance.
Biochemical and biophysical research communications (1990-01-30)
K Tajima, M Shigematsu, J Jinno, Y Kawano, K Mikami, K Ishizu, H Ohya-Nishiguchi, M Shigamatsu
ABSTRACT
Coordination reaction between linolenic-acid-hydroperoxide (LHPO) and chloro(5, 10, 15, 20-tetraphenyl)-porphyrinato iron (III), Fe(III)TPPCl, was investigated by means of ESR. ESR spectra of the ferric low-spin complex (g1 = 2.336, g2 = 2.174 and g3 = 1.929) was recorded for the mixture prepared by mixing Fe(III)TPPCl and LHPO at -78 degrees C in the presence of alkaline reagent. ESR line width of complex was broadened when 17O2 labeled LHPO was used for ESR measurement. In terms of the g-parameters of the ferric low-spin species, this complex was concluded to be Fe(III)TPP(-OCH3)(-OO-linolenic acid) type complex.
Sigma-Aldrich
5,10,15,20-Tetraphenyl-21H,23H-porphine iron(III) chloride, ≥94% (HPLC)