Merck
  • Dietary curcumin and limonin suppress CD4+ T-cell proliferation and interleukin-2 production in mice.
Dietary curcumin and limonin suppress CD4+ T-cell proliferation and interleukin-2 production in mice.
The Journal of nutrition (2009-03-27)
Wooki Kim, Yang-Yi Fan, Roger Smith, Bhimanagouda Patil, Guddadarangavvanahally K Jayaprakasha, David N McMurray, Robert S Chapkin
ABSTRACT
Phytochemicals may reduce chronic inflammation and cancer risk in part by modulating T-cell nuclear factor-kappaB (NF-kappaB) activation. Therefore, we examined the effects of curcumin (Cur) and limonin (Lim) feeding on NF-kappaB-dependent CD4(+) T-cell proliferation. DO11.10 transgenic mice (n = 5-7) were fed diets containing 1% Cur or 0.02% Lim combined with either (n-6) PUFA [5% corn oil (CO)] or (n-3) PUFA [4% fish oil+1% corn oil (FO)] for 2 wk, followed by splenic CD4(+) T-cell isolation and stimulation with ovalbumin peptide 323-339 (OVA) and antigen-presenting cells from mice fed a conventional nonpurified rodent diet. Both Cur and Lim diets suppressed (P < 0.05) NF-kappaB p65 nuclear translocation in activated CD4(+) T-cells. In contrast, activator protein-1 (c-Jun) and nuclear factor of activated T-cells c1 were not affected compared with the CO control diet (no Cur or Lim). CD4(+) T-cell proliferation in response to either mitogenic anti-CD3/28 monoclonal antibodies (mAb) or antigenic stimulation by OVA was also suppressed (P < 0.05) by Cur as assessed by carboxyfluorescein succinimidyl ester staining. In contrast, interleukin-2 production was not directly associated with NF-kappaB status. Interestingly, dietary combination with FO enhanced the suppressive effects (P < 0.05) of Cur or Lim with respect to CD4(+) T-cell proliferation in response to anti-CD3/28 mAb. These results suggest that combination chemotherapy (FO+Cur or Lim) may favorably modulate CD4(+) T-cell-mediated inflammation.
Sigma-Aldrich
Limonin, from citrus seeds, >90% (HPLC)