Merck
  • Side reactions in the pyroglutamic acid-7-amino-4-methylcoumarin synthesis.
Side reactions in the pyroglutamic acid-7-amino-4-methylcoumarin synthesis.
International journal of peptide and protein research (1996-07-01)
D Zicane, M Petrova, Z Tetere, G Kreishman, E Brooks, J Karlson, U Kalejs
ABSTRACT
N-Formylpyroglutamic acid-7-amido-4-methylcoumarine and pyroglutamyl-pyroglutamic acid-7-amido-4-methylcoumarin are the major products in the synthesis of pyroglutamic acid-7-amido-4-methylcoumarin by phosphorus pentachloride in dimethylformamide and dicyclohexylcarbodiimide under pyridine activation.
Sigma-Aldrich
Phosphorus trichloride, ReagentPlus®, 99%
Sigma-Aldrich
Phosphorus trichloride, 99.999% trace metals basis