Merck
  • The hydroxylation, dechlorination, and glucuronidation of 4,4'-dichlorobiphenyl (4-DCB) by human hepatic microsomes.
The hydroxylation, dechlorination, and glucuronidation of 4,4'-dichlorobiphenyl (4-DCB) by human hepatic microsomes.
Biochemical pharmacology (1984-11-01)
R G Schnellmann, R F Volp, C W Putnam, I G Sipes
ABSTRACT
Since chlorine placement and the degree of chlorination of the biphenyl nucleus play an important role in the metabolism and ultimate elimination of polychlorinated biphenyls (PCBs), we have studied the metabolism of 4,4'-dichlorobiphenyl (4-DCB) by human hepatic microsomes. This low molecular weight PCB congener is substituted at the preferred site of metabolism (para-position). 4-DCB was metabolized by human microsomes with a Km of 0.43 microM and a Vmax of 1.2 pmoles/mg microsomal protein/min. Six metabolites were identified: 4,4'-dichloro-3,3'-biphenyldiol, 4'-chloro-3-biphenylol, 4'-chloro-4-biphenylol, 4,4'-dichloro-2-biphenylol, 4,4'-dichloro-3-biphenylol (most abundant), and 3,4'-dichloro-4-biphenylol. [14C]-4-DCB equivalents were found to covalently bind to microsomal protein. Addition of a 1 mM concentration of reduced glutathione decreased the degree of covalent binding. These data suggest that human microsomes metabolize this PCB through an arene oxide and that an "NIH shift" occurs. When UDPGA was added to the incubation, human microsomal glucuronosyltransferase catalyzed the formation of the glucuronide of the major metabolite, 4,4'-dichloro-3-biphenylol. These and previous in vitro results show that the biotransformation of PCBs by humans is governed by the same principles established for the in vivo biotransformation of PCBs by the rat, mouse and monkey. That is, PCBs without two adjacent unsubstituted carbon atoms are poorly metabolized and that an unsubstituted para-position facilitates metabolism.
Product Description
Supelco
PCB No 15, analytical standard
Pricing and availability is not currently available.