Merck
  • Spatial and temporal variations in pentachlorophenol dissipation at the aerobic--anaerobic interfaces of flooded paddy soils.
Spatial and temporal variations in pentachlorophenol dissipation at the aerobic--anaerobic interfaces of flooded paddy soils.
Environmental pollution (Barking, Essex : 1987) (2013-05-01)
Jiajiang Lin, Yan Xu, Philip C Brookes, Yan He, Jianming Xu
ABSTRACT
Pentachlorophenol (PCP) dissipation occurs naturally in flooded soils and although dissipation half-lives vary between soil profiles at the millimeter-scale the reason is poorly understood. Vertical variations of PCP dissipation were investigated in three typical Chinese paddy soils; Soil 1 (Umbraqualf), Soil 2 (Plinthudult) and Soil 3 (Tropudult). The soil depth was divided into a surface and a deep layer based upon different PCP dissipations in the surface layer of 40-93, 42-88 and 16-100% for Soils 1-3 respectively. In the deep layer, PCP was greatly dissipated in Soil 2, but much less in Soil 1 and Soil 3. Correlation analysis indicated that SO4(2-) and Fe(III) were negatively related to PCP dissipation. SO4(2-) and Cl(-) were highly mobile in the flooded soil profiles. Fe(III) reduction increased with increasing soil depth, and was inhibited by high SO4(2-) concentrations.
Product Description
Supelco
Pentachlorophenol solution, certified reference material, 5000 μg/mL in methanol