Merck
  • Asymmetric one-pot sequential Mannich/hydroamination reaction by organo- and gold catalysts: synthesis of spiro[pyrrolidin-3,2'-oxindole] derivatives.
Asymmetric one-pot sequential Mannich/hydroamination reaction by organo- and gold catalysts: synthesis of spiro[pyrrolidin-3,2'-oxindole] derivatives.
Organic letters (2013-04-09)
Xianjie Chen, Hui Chen, Xun Ji, Hualiang Jiang, Zhu-Jun Yao, Hong Liu
ABSTRACT
An asymmetric organo- and gold-catalyzed one-pot sequential Mannich/hydroamination reaction has been developed. Using this protocol, spiro[pyrrolidin-3,2'-oxindole] derivatives were synthesized in good yields (up to 91%) and excellent enantioselectivities (up to 97% ee).
Sigma-Aldrich
2-Oxindole, 97%