Merck
  • Chronic food administration of Salvia sclarea oil reduces animals' anxious and dominant behavior.
Chronic food administration of Salvia sclarea oil reduces animals' anxious and dominant behavior.
Journal of medicinal food (2013-03-01)
Moshe Gross, Elimelech Nesher, Tatiana Tikhonov, Olga Raz, Albert Pinhasov
ABSTRACT
Recent studies indicate that an oil extract from Salvia sclarea may provide clinical benefits in various pathological conditions. In comparison to extracts from other Salvia species, S. sclarea oil contains twice as much omega-3 fatty acids, which are involved in eicosanoid synthesis pathways, and has been found to contain significant levels of the psychoactive monoterpane linalool. In the present study, we examined the mood stabilizing and anxiolytic-like effects of chronic food administration of S. sclarea oil extract on behavioral and physiological parameters of mice with prominent dominant and submissive features in behavioral assays used to test mood stabilizing and antidepressant drugs. Experimental animals received oil supplemented food from the age of 4 weeks or from conception via their pregnant dams. Each age group received either S. sclarea oil- or sunflower oil-enriched feed. Dominant animals, whose pregnant mothers received S. sclarea oil-enriched feed from the date of conception, showed a significant reduction of dominant and anxiety-like behavior, in comparison to their sunflower oil-treated counterparts. S. sclarea oil-treated submissive animals exhibited a similar tendency, and showed a significant reduction in blood corticosterone levels. These findings enforce the hypothesis that S. sclarea oil possesses anxiolytic properties.
Product Description
Supelco
Sunflower seed oil, analytical standard