Merck
  • Fluorescent probe for biological gas SO2 derivatives bisulfite and sulfite.
Fluorescent probe for biological gas SO2 derivatives bisulfite and sulfite.
Chemical communications (Cambridge, England) (2013-02-26)
Yuan-Qiang Sun, Jing Liu, Jingyu Zhang, Ting Yang, Wei Guo
ABSTRACT
A coumarin-hemicyanine dye was reported for ratiometric fluorescent detection of SO2 derivatives HSO3(-) and SO3(2-) based on a novel addition-rearrangement cascade reaction.
Sigma-Aldrich
Sulfur dioxide, ≥99.9%