Merck
  • Photoconductive properties of a π-conjugated α-cyclodextrin containing [2]rotaxane and its corresponding molecular dumbbell.
Photoconductive properties of a π-conjugated α-cyclodextrin containing [2]rotaxane and its corresponding molecular dumbbell.
Physical chemistry chemical physics : PCCP (2013-01-31)
Ioanna Deligkiozi, Raffaello Papadakis, Athanase Tsolomitis
ABSTRACT
The photoconductive properties of an azo-containing [2]rotaxane, bearing a π-conjugated axial part and its corresponding dumbbell compound are investigated. Structural effects on the observed photoconductive behavior in both cases are discussed. The photoresponsive behavior of the title [2]rotaxane was proved to be more intense than that of its analogue lacking the α-cyclodextrin (α-CyD) macrocycle. A mechanism of the photoinduced charge transport in both cases is proposed.
Sigma-Aldrich
α-Cyclodextrin, powder, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
α-Cyclodextrin, ≥98%
Sigma-Aldrich
α-Cyclodextrin, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0% (HPLC)
Sigma-Aldrich
α-Cyclodextrin, purum, ≥98.0% (HPLC)