Merck
  • Analysis of UV and vibrational spectra (FT-IR and FT-Raman) of hexachlorocyclotriphosphazene based on normal coordinate analysis, MP2 and DFT calculations.
Analysis of UV and vibrational spectra (FT-IR and FT-Raman) of hexachlorocyclotriphosphazene based on normal coordinate analysis, MP2 and DFT calculations.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2013-01-26)
Wajdi M Zoghaib, John Husband, Usama A Soliman, Ibrahim A Shaaban, Tarek A Mohamed
ABSTRACT
The Raman (1400-100 cm(-1)) and infrared (4000-400 cm(-1)) of solid hexachlorocyclotriphosphazene, P(3)N(3)Cl(6) (HCCTP) were recorded. The conformational energies were calculated using MP2 and DFT (B3LYP and B3PW91) methods utilizing a variety of basis sets up to 6-311+G(d). On the basis of D(3h) symmetry, the simulated vibrational spectra of P(3)N(3)Cl(6) from MP2 and DFT methods were in excellent agreement with those obtained experimentally. Additionally, Frontier Molecular Orbitals and electronic transitions were predicted using steady state and time dependent DFT(B3LYP)/PCM calculations respectively, each employing the 6-311+G(d,p) optimized structural parameters. The predicted wavelengths were in excellent agreement with experimental values when CH(2)Cl(2) was used as solvent. The (14)N and (31)P chemical shifts were predicted with B3LYP/6-311+G(2d,p) calculations using the GIAO technique with solvent effect modeled using the PCM method. The computed structural parameters of the planar P(3)N(3)Cl(6) (D(3h)) agree well with experimental values from both X-ray and electron diffraction data with slight distortions observed due to lattice defects in the solid phase. The experimental/computational results favor a slightly distorted D(3h) symmetry for the title compound in the gas and solid phases and in solution (τPNPN and τNPNP ranged from 0.018° to 0.90°). Aided by normal coordinate analysis, and the simulated vibrational spectra utilizing MP2, B3LYP and B3PW91 methods at 6-31G(d) basis set, revised and complete vibrational assignments for all fundamentals are provided herein.
Sigma-Aldrich
Phosphonitrilic chloride trimer, 99%