Merck
  • TiN nanoparticles on CNT-graphene hybrid support as noble-metal-free counter electrode for quantum-dot-sensitized solar cells.
TiN nanoparticles on CNT-graphene hybrid support as noble-metal-free counter electrode for quantum-dot-sensitized solar cells.
ChemSusChem (2013-01-11)
Duck Hyun Youn, Minsu Seol, Jae Young Kim, Ji-Wook Jang, Youngwoo Choi, Kijung Yong, Jae Sung Lee
ABSTRACT
The development of an efficient noble-metal-free counter electrode is crucial for possible applications of quantum-dot-sensitized solar cells (QDSSCs). Herein, we present TiN nanoparticles on a carbon nanotube (CNT)-graphene hybrid support as a noble-metal-free counter electrode for QDSSCs employing a polysulfide electrolyte. The resulting TiN/CNT-graphene possesses an extremely high surface roughness, a good metal-support interaction, and less aggregation relative to unsupported TiN; it also has superior solar power conversion efficiency (4.13 %) when applying a metal mask, which is much higher than that of the state-of-the-art Au electrode (3.35 %). Based on electrochemical impedance spectroscopy measurements, the enhancement is ascribed to a synergistic effect between TiN nanoparticles and the CNT-graphene hybrid, the roles of which are to provide active sites for the reduction of polysulfide ions and electron pathways to TiN nanoparticles, respectively. The combination of graphene and CNTs leads to a favorable morphology that prevents stacking of graphene or bundling of CNTs, which maximizes the contact of the support with TiN nanoparticles and improves electron-transfer capability relative to either carbon material alone.
Sigma-Aldrich
Titanium nitride, <3 μm