Merck
  • Astrocytes regulate myelin clearance through recruitment of microglia during cuprizone-induced demyelination.
Astrocytes regulate myelin clearance through recruitment of microglia during cuprizone-induced demyelination.
Brain : a journal of neurology (2012-12-26)
Thomas Skripuletz, Diane Hackstette, Katharina Bauer, Viktoria Gudi, Refik Pul, Elke Voss, Katharina Berger, Markus Kipp, Wolfgang Baumgärtner, Martin Stangel
ABSTRACT
Recent evidence suggests that astrocytes play an important role in regulating de- and remyelination in multiple sclerosis. The role of astrocytes is controversial, and both beneficial as well as detrimental effects are being discussed. We performed loss-of-function studies based on astrocyte depletion in a cuprizone-induced rodent model of demyelination. This led to strong astrogliosis accompanied by microgliosis and demyelination in C57BL/6 wild-type mice. Ablation of astrocytes in glial fibrillary acidic protein-thymidine kinase transgenic mice was associated with a failure of damaged myelin removal and a consecutive delay in remyelination. Despite oligodendrocyte death, myelin was still present, but ultrastructual investigations showed that the myelin structure was loosened and this damaged myelin did not protect axons. These alterations were associated with a decrease in microglial activation. Thus, our results show that astrocyte loss does not prevent myelin damage, but clearance of damaged myelin through recruitment of microglia is impaired. Further studies suggest that this process is regulated by the chemokine CXCL10. As a consequence of the delayed removal of myelin debris, remyelination and oligodendrocyte precursor cell proliferation were impaired. Experiments omitting the influence of myelin debris demonstrated an additional beneficial effect of astrocytes on oligodendrocyte regeneration during remyelination. In conclusion, these data demonstrate for the first time in vivo that astrocytes provide the signal environment that forms the basis for the recruitment of microglia to clear myelin debris, a process required for subsequent repair mechanisms. This is of great importance to understanding regenerative processes in demyelinating diseases such as multiple sclerosis.
Product Description
Supelco
Bis(cyclohexanone)oxaldihydrazone, for spectrophotometric det. of Cu, ≥99.0%
Sigma-Aldrich
Bis(cyclohexanone)oxaldihydrazone, ≥95% (TLC)