Merck
Antibacterial drug leads targeting isoprenoid biosynthesis.
Proceedings of the National Academy of Sciences of the United States of America (2012-12-19)
Wei Zhu, Yonghui Zhang, William Sinko, Mary E Hensler, Joshua Olson, Katie J Molohon, Steffen Lindert, Rong Cao, Kai Li, Ke Wang, Yang Wang, Yi-Liang Liu, Anna Sankovsky, César Augusto F de Oliveira, Douglas A Mitchell, Victor Nizet, J Andrew McCammon, Eric Oldfield
ABSTRACT
With the rise in resistance to antibiotics such as methicillin, there is a need for new drugs. We report here the discovery and X-ray crystallographic structures of 10 chemically diverse compounds (benzoic, diketo, and phosphonic acids, as well as a bisamidine and a bisamine) that inhibit bacterial undecaprenyl diphosphate synthase, an essential enzyme involved in cell wall biosynthesis. The inhibitors bind to one or more of the four undecaprenyl diphosphate synthase inhibitor binding sites identified previously, with the most active leads binding to site 4, outside the catalytic center. The most potent leads are active against Staphylococcus aureus [minimal inhibitory concentration (MIC)(90) ∼0.25 µg/mL], and one potently synergizes with methicillin (fractional inhibitory concentration index = 0.25) and is protective in a mouse infection model. These results provide numerous leads for antibacterial development and open up the possibility of restoring sensitivity to drugs such as methicillin, using combination therapies.
Product Description
Millipore
MRSA Selective Supplement, suitable for microbiology
Sigma-Aldrich
Methicillin sodium salt, ≥95% (HPLC)