Merck
  • INF-β1b therapy modulates L-arginine and nitric oxide metabolism in patients with relapse remittent multiple sclerosis.
INF-β1b therapy modulates L-arginine and nitric oxide metabolism in patients with relapse remittent multiple sclerosis.
Journal of the neurological sciences (2012-10-03)
Ivana Stojanovic, Slobodan Vojinovic, Srdjan Ljubisavljevic, Radmila Pavlovic, Jelena Basic, Dusica Pavlovic, Andjelka Ilic, Tatjana Cvetkovic, Maja Stukalov
ABSTRACT
The scope of this study is the examination of NO(2)+NO(3), 3-nitrotyrosine (3-NT), S-nitrosothiols (RSNO), arginase activity and asymmetric (ADMA) and symmetric (SDMA) dimethyl-L-arginine concentrations in plasma of MS patients during interferon-β1b therapy. The study population included 15 (12 women, 3 men) untreated MS patients and 12 (10 women, 2 men) interferon-β1b treated MS patients with clinically definite relapsing MS (McDonalds criteria) for at least 1 year and a baseline EDSS score of 1.0 to 3.5 inclusive. Patients were treated with 250 μg IU interferon-β1b s.c. every second day during 30 months. The disease course was evaluated using correlations between baseline EDSS score and relapse rates in both groups. During interferon-β1b treatment, EDSS scores in treated patients were decreased compared to untreated ones - after 18 and 30 months (p<0.05). In interferon-β1b treated MS patients, NO(2)+NO(3), 3-NT and RSNO plasma concentrations were significantly lower (p<0.05), while arginase activity, ADMA and SDMA levels were significantly increased (p<0.05) during the therapy, compared to the baseline levels in treated patients. The investigated parameters may be the new biomarkers, providing information for the therapeutic approach and valuable in clinical monitoring.
Sigma-Aldrich
NG,NG′-Dimethyl-L-arginine di(p-hydroxyazobenzene-p′-sulfonate) salt, ≥99% (TLC)