Merck
  • Molecular interaction fields based descriptors to interpret and compare chromatographic indexes.
Molecular interaction fields based descriptors to interpret and compare chromatographic indexes.
Journal of chromatography. A (2012-07-17)
G Ermondi, G Caron
ABSTRACT
Molecular Interaction Fields (MIFs) based descriptors can be conveniently used to characterize and compare chromatographic scales. In this study, Quantitative Structure-Retention Relationships (QSRR) for eight different chromatographic systems were obtained with VolSurf+ descriptors and Partial Least Squares (PLS). A new and purpose-designed analysis tool highlights the different balance of intermolecular interactions governing solute retention, and estimates the similarity between chromatographic systems.
Sigma-Aldrich
3-Amino-1,2,4-triazole, ≥95% (TLC)