Merck
  • Safrole induces G0/G1 phase arrest via inhibition of cyclin E and provokes apoptosis through endoplasmic reticulum stress and mitochondrion-dependent pathways in human leukemia HL-60 cells.
Safrole induces G0/G1 phase arrest via inhibition of cyclin E and provokes apoptosis through endoplasmic reticulum stress and mitochondrion-dependent pathways in human leukemia HL-60 cells.
Anticancer research (2012-05-18)
Chun-Shu Yu, An-Cheng Huang, Jai-Sing Yang, Chien-Chih Yu, Chin-Chung Lin, Hsiung-Kwang Chung, Yi-Ping Huang, Fu-Shin Chueh, Jing-Gung Chung
ABSTRACT
Safrole, a component of Piper betle inflorescence, is a carcinogen which has been demonstrated to induce apoptosis on human oral cancer HSC-3 cells in vitro and to inhibit HSC-3 cells in xenograft tumor cells in vivo. In our previous study, safrole promoted phagocytosis by macrophages and natural killer cell cytotoxicity in normal BALB/c mice. The cytotoxic effects of safrole on HL-60 cells were investigated by using flow cytometric analysis, comet assay, 4',6-diamidino-2-phenylindole (DAPI) staining, western blotting and confocal laser microscopy. The obtained results indicate that safrole induced a cytotoxic response through reducing the percentage of viable cells and induction of apoptosis in HL-60 cells in a dose-dependent manner. DAPI staining and comet assay also showed that safrole induced apoptosis (chromatin condensation) and DNA damage in HL-60 cells. The flow cytometric assay showed that safrole increased the production of reactive oxygen species (ROS) and Ca(2+) and reduced the mitochondrial membrane potential in HL-60 cells. Safrole enhanced the levels of the pro-apoptotic protein BAX, inhibited those of the anti-apoptotic protein BCL-2 and promoted the levels of apoptosis-inducing factor (AIF) and endonuclease G (Endo G) in HL-60 cells. Furthermore, safrole promoted the expression of glucose-regulated protein 78 (GRP78), growth arrest- and DNA damage-inducible gene 153 (GADD153) and of activating transcription factor 6α (ATF-6α). Based on these findings, we suggest that safrole-induced apoptosis in HL-60 cells is mediated through the ER stress and intrinsic signaling pathways.
Sigma-Aldrich
Safrole, ≥97%