Merck
  • Fluoride-mediated elimination of allyl sulfones: application to the synthesis of a 2,4-dimethyl-A-ring vitamin D3 analogue.
Fluoride-mediated elimination of allyl sulfones: application to the synthesis of a 2,4-dimethyl-A-ring vitamin D3 analogue.
The Journal of organic chemistry (2012-04-27)
Vikas Sikervar, James C Fleet, Philip L Fuchs
ABSTRACT
A coupling strategy for the synthesis of 2,4-dimethyl-1α,25(OH)(2)D(3) is achieved which involves methylation of a pro-A ring vinyl sulfone and in situ traping of the allyl sulfonyl anion with a CD ring allyl chloride. TBAF-promoted 1,2-eliminative desulfonylation and concomitant silyl ether deprotection gives the vitamin D(3) analogue.
Sigma-Aldrich
Allyl chloride, ReagentPlus®, 99%
SAFC
Allyl chloride
Sigma-Aldrich
Allyl chloride, reagent grade, 98%