Merck
  • Experimental and DFT theoretical studies of surface enhanced Raman scattering effect on the silver nano arrays modified electrode.
Experimental and DFT theoretical studies of surface enhanced Raman scattering effect on the silver nano arrays modified electrode.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-04-13)
Lisheng Zhang, Yan Fang, Peijie Wang
ABSTRACT
Well-ordered silver nano arrays, prepared using anodic aluminum oxide (AAO) template by laser molecular beam epitaxy (L-MBE) method, were adhered to the work electrode using conductive adhesive to be a Surface-enhanced Raman scattering (SERS) substrate. Variable SERS signals of paranitrobenzoic acid (PNA) on the electrode modified with silver nano arrays were recorded with electric potential ranging from 0.1 to -0.5 V. The SERS spectra of PNA using DFT-B3PW91 with lanl2dz based on two models were calculated. It indicate that the adsorption orientation of probe molecules on the silver nano arrays with potential change of the electrode.
Sigma-Aldrich
4-Nitrobenzoic acid, 98%
Sigma-Aldrich
4-Nitrobenzoic acid, purum, ≥98.0% (HPLC)