Merck
  • Tank mixture additives approach to improve efficiency of bentazon against broadleaf weeds in peas.
Tank mixture additives approach to improve efficiency of bentazon against broadleaf weeds in peas.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2012-03-20)
Mohamed A Balah, Ahmad Hanafi, Sherif B Abdel Ghani
ABSTRACT
Efficiency of different tank-mixed additives with bentazon at half rate was investigated on (Malva parviflora) and other broad leaf weeds compared with bentazon at the full recommended rate without additives in peas in open field. All the tested additives enhanced the efficiency of bentazon at the half rate. Nonyl phenol and toximol S proved to be the most effective additives in comparison with the full rate treatment. The tested treatments did not show any significant effect on chlorophyll content and soil microorganisms. Bentazon residues were determined in certain treatments to investigate the effect of the tested additives on bentazon deposition. Samples were extracted using QuEChERS method and residues were determined using LC-MS/MS. Residues after 24 hours in the half rate treatment reached 4 times lower than the Maximum Residues Limit (MRL) (0.11 mg kg(-1)), compared to the full rate treatment (0.51 mg kg(-1)), that was slightly above the MRL.