Merck
  • Changes in fibrinopeptide A peptides in the sera of rats chronically exposed to low doses of 2,3,7,8-tetrachlorodibenzo-p-dioxin (TCDD).
Changes in fibrinopeptide A peptides in the sera of rats chronically exposed to low doses of 2,3,7,8-tetrachlorodibenzo-p-dioxin (TCDD).
Environmental toxicology and pharmacology (2012-01-10)
Xi Chen, Shi-wei Ma, Xiao-ming Ma, Yuan-jing Xu, Nai-jun Tang
ABSTRACT
2,3,7,8-Tetrachlorodibenzo-p-dioxin (TCDD) is a ubiquitously distributed endocrine disruptors. To investigate peptide changes in the sera of rats chronically exposed to TCDD and to explore the association of these changes with liver morphology, TCDD was administrated to male rats at doses of 140, 350, and 875 ng/kg/week for 29 weeks. Serum was collected and proteomic analysis was performed using automated Bruker Daltonics ClinProt with matrix-assisted laser desorption/ionization time-of-flight mass spectrometer. One peptide at 1740.89 was found to be significantly decreased and further identified with nano LC-MS/MS system. The MS BLAST homology search engine reported the peptide to be a partial sequence of fibrinopeptide A. Liver fatty degeneration and necrosis were assessed by hematoxylin and eosin staining. Liver fatty degeneration and necrosis were both found to be significantly increased after TCDD exposure. Levels of fibrinopeptide A were significantly correlated with liver fatty degeneration and necrosis.
Sigma-Aldrich
Fibrinopeptide A human, ≥96% (HPLC)