Merck
  • Evaluating auxin distribution in tomato (Solanum lycopersicum) through an analysis of the PIN and AUX/LAX gene families.
Evaluating auxin distribution in tomato (Solanum lycopersicum) through an analysis of the PIN and AUX/LAX gene families.
The Plant journal : for cell and molecular biology (2012-01-04)
Richard J Pattison, Carmen Catalá
ABSTRACT
The temporal and spatial control of auxin distribution has a key role in the regulation of plant growth and development, and much has been learnt about the mechanisms that influence auxin pools and gradients in vegetative tissues, particularly in Arabidopsis. For example polar auxin transport, mediated by PIN and AUX/LAX proteins, is central to the control of auxin distribution. In contrast, very little information is known about the dynamics of auxin distribution and the molecular basis of its transport within and between fruit tissues, despite the fact that auxin regulates many aspects of fruit development, which include fruit formation, expansion, ripening and abscission. In addition, functional information regarding the key regulators of auxin fluxes during both vegetative and reproductive development in species other than Arabidopsis is scarce. To address these issues, we have investigated the spatiotemporal distribution of auxin during tomato (Solanum lycopersicum) fruit development and the function of the PIN and AUX/LAX gene families. Differential concentrations of auxin become apparent during early fruit growth, with auxin levels being higher in internal tissues than in the fruit pericarp and the pattern of auxin accumulation depended on polar transport. Ten tomato PIN (SlPIN1 to 10) and five AUX/LAX (SlLAX1 to 5) genes were identified and found to display heterogeneous expression patterns, with tissue and developmental-stage specificity. RNAi-mediated co-silencing of SlPIN4 and SlPIN3 did not affect fruit development, which suggested functional redundancy of PIN proteins, but did lead to a vegetative phenotype, and revealed a role for these genes in the regulation of tomato shoot architecture.