Merck
  • A proposal of interpretive criteria for cefoperazone applicable to bovine mastitis pathogens.
A proposal of interpretive criteria for cefoperazone applicable to bovine mastitis pathogens.
Veterinary microbiology (2011-12-23)
Andrea T Fessler, Heike Kaspar, Cynthia J Lindeman, Michael R Stegemann, Thomas Peters, Joachim Mankertz, Jeffrey L Watts, Stefan Schwarz
ABSTRACT
The correct assessment of mastitis pathogens for their susceptibility/resistance to cefoperazone is currently hampered by the lack of harmonized test conditions and interpretive criteria. The aim of this study was to provide a proposal for clinical breakpoints of cefoperazone which are applicable to Staphylococcus aureus, coagulase-negative staphylococci, Escherichia coli, Streptococcus agalactiae, Streptococcus dysgalactiae and Streptococcus uberis from cases of bovine mastitis and better reflect the situation in the bovine udder than breakpoints adopted from human medicine. For this, pharmacological data and clinical efficacy data of the documents submitted for approval of cefoperazone have been revisited. In addition, 1086 bacterial pathogens of the aforementioned six species/groups collected in Germany and in the USA during recent years were tested in parallel for their cefoperazone MICs and the zone diameters using a 75 μg disk. Subsequently, MICs were plotted against zone diameters. Based on the pharmacological data, the clinical efficacy and the microbiological data, a proposal was made for veterinary-specific breakpoints which classify members of the aforementioned species/groups as (a) susceptible to cefoperazone when their MIC is ≤ 2 μg/ml and their zone diameters are ≥ 27 mm (staphylococci or E. coli) or ≥ 21 mm (streptococci), (b) intermediate when their MIC is 4 μg/ml and their zone diameters are 22-26 mm (staphylococci or E. coli) or 16-20mm (streptococci), and (c) resistant when their MIC is ≥ 8 μg/ml and their zone diameters are ≤ 21 mm (staphylococci or E. coli) or ≤ 15 mm (streptococci).
Sigma-Aldrich
Cefoperazone sodium salt, 870 - 1015 μg/mg anhydrous basis