Merck
  • Genotoxic and cytotoxic evaluation of the herbicide flurochloridone on Chinese hamster ovary (CHO-K1) cells.
Genotoxic and cytotoxic evaluation of the herbicide flurochloridone on Chinese hamster ovary (CHO-K1) cells.
Toxicology in vitro : an international journal published in association with BIBRA (2011-11-15)
Noelia Nikoloff, Sonia Soloneski, Marcelo L Larramendy
ABSTRACT
The in vitro effects of flurochloridone (FLC) and its formulations Twin Gold PackĀ® (25% a.i.) and RainbowĀ® (25% a.i.) were evaluated on Chinese hamster ovary (CHO-K1) cells by genotoxicity [sister chromatid exchange (SCE)] and cytotoxicity [cell-cycle progression, proliferative rate index (PRI), mitotic index (MI), MTT, and neutral red] end points. Cells were treated for 24h within the 0.25-15Ī¼g/ml concentration range. FLC and Twin Pack GoldĀ® induced a significant and equivalent increase in SCEs regardless of the concentration. RainbowĀ®-induced SCEs at concentrations higher than 2.5Ī¼g/ml; however, the increases were always lower than those induced by FLC and Twin Pack GoldĀ®. For all compounds, the PRI decreased as a function of the concentration titrated into cultures. Whereas only the highest FLC and Twin Pack GoldĀ® concentrations induced a significant reduction of the MI, all tested RainbowĀ® concentrations induced MI inhibition. Overall, the results demonstrated that although all compounds were not able to reduce the lysosomal activity, the mitochondrial activity was diminished when the highest concentrations were employed. These observations represent the first study analyzing the genotoxic and cytotoxic effects exerted by FLC and two formulated products on mammalian cells in vitro, at least on CHO-K1 cells.
Product Description
Supelco
Fluridon, PESTANALĀ®, analytical standard