Merck
  • Determination of 4(5)-methylimidazole in soy sauce and other foods by LC-MS/MS after solid-phase extraction.
Determination of 4(5)-methylimidazole in soy sauce and other foods by LC-MS/MS after solid-phase extraction.
Journal of agricultural and food chemistry (2011-09-02)
Hitomi Yamaguchi, Tsutomu Masuda
ABSTRACT
A method for the determination of 4(5)-methylimidazole (4MeI) in naturally brewed soy sauce was developed for the first time using solid-phase extraction (SPE) and liquid chromatography-tandem mass spectrometry (LC-MS/MS). SPE on silica-based reversed-phase cartridges with heptafluorobutyric acid as an ion-pairing reagent was used for the efficient cleanup of 4MeI. A multimode ODS column was employed for the chromatographic separation. To subtract the matrix effect during LC-MS/MS analysis, a standard addition method was used. The levels of 4MeI found in naturally brewed soy sauce were extremely low (ranging from <0.002 to 0.023 μg/g), whereas those in soy sauces containing caramel color were generally high (ranging from 0.43 to 4.8 μg/g). The method proved to be useful for the analysis of 4MeI in other foods such as caramel colors, drinks, and Worcestershire sauce.
Sigma-Aldrich
4(5)-Methylimidazole, 98%
Pricing and availability is not currently available.