Merck
  • Synthesis and Diels-Alder cycloadditions of exo-imidazolidin-2-one dienes.
Synthesis and Diels-Alder cycloadditions of exo-imidazolidin-2-one dienes.
The Journal of organic chemistry (2011-08-17)
Rafael Bautista, Pablo Bernal, Rafael Herrera, Blanca M Santoyo, J Miguel Lazcano-Seres, Francisco Delgado, Joaquín Tamariz
ABSTRACT
An efficient and versatile synthesis of novel exo-imidazolidin-2-one dienes is described. This involves the base-assisted condensation/cyclization cascade reaction of the monoimino derivatives of diacetyl with a series of isocyanates. This methodology enables preparation of symmetrical dienes, as long as the substrates have the same N substituent. Moreover, use of different N-substituted starting materials leads to formation of nonsymmetrical dienes. The reactivity of these dienes was evaluated in Diels-Alder reactions, showing a high reactivity.
Sigma-Aldrich
2-Imidazolidone, 96%