Merck
  • Production of anticancer polyenes through precursor-directed biosynthesis.
Production of anticancer polyenes through precursor-directed biosynthesis.
Organic & biomolecular chemistry (2011-07-28)
Benjamin R Clark, Stephen O'Connor, Deirdre Fox, Jacques Leroy, Cormac D Murphy
ABSTRACT
The biosynthesis of the pyrrolyl moiety of the fungal metabolite rumbrin originates from pyrrole-2-carboxylic acid. In an effort to produce novel derivatives with enhanced biological activity a series of substituted pyrrole-2-carboxylates were synthesised and incubated with the producing organism, Auxarthron umbrinum. Several 4-halo-pyrrole-2-carboxylic acids were incorporated into the metabolite yielding three new derivatives: 3-fluoro-, 3-chloro- and 3-bromo-isorumbrin, which were generated in milligram quantities enabling cytotoxicity assays to be conducted. The 3-chloro- and 3-bromo-isorumbrins had improved activity against HeLa cells compared with rumbrin; 3-bromoisorumbrin also showed dramatically improved activity towards a lung cancer cell line (A549).
Sigma-Aldrich
Pyrrole-2-carboxylic acid, 99%