Merck
  • Stability of heterochiral hybrid membrane made of bacterial sn-G3P lipids and archaeal sn-G1P lipids.
Stability of heterochiral hybrid membrane made of bacterial sn-G3P lipids and archaeal sn-G1P lipids.
Biochemistry (2011-04-09)
Haruo Shimada, Akihiko Yamagishi
ABSTRACT
The structure of membrane lipids in Archaea is different from those of Bacteria and Eucarya in many ways including the chirality of the glycerol backbone. Until now, heterochiral membranes were believed to be unstable; thus, no cellular organism could have existed before the separation of the groups of life. In this study, we tested the formation of heterochiral hybrid membrane made of Bacterial sn-glycerol-3-phosphate-type polar lipid and Archaeal sn-glycerol-1-phosphate-type polar lipid using the fluorescence probe. The stability of the hybrid liposomes made of phosphatidylethanolamines or phosphatidylcholines or polar lipids of thermophilic Bacteria and polar lipids of Archaea were investigated. The hybrid liposomes are all stable compared with homochiral liposome made of dimyristoylphosphatidylethanolamine and dipalmitoylphosphatidylcholine. However, the stability was drastically changed with increasing carbon chain length. Accordingly, "chirality" may not be but chain length is important. From these results, we suggest that the heterochiral hybrid membrane could be used as the membrane lipid for the last universal common ancestor (Commonote) before the emergence of Archaea and Bacteria.
Sigma-Aldrich
DL-α-Glycerol phosphate magnesium salt hydrate, ~85% (KT)