Merck
  • Accumulation and residue of napropamide in alfalfa (Medicago sativa) and soil involved in toxic response.
Accumulation and residue of napropamide in alfalfa (Medicago sativa) and soil involved in toxic response.
Journal of hazardous materials (2011-03-29)
Li E Cui, Hong Yang
ABSTRACT
Napropamide belongs to the amide herbicide family and widely used to control weeds in farmland. Intensive use of the herbicide has resulted in widespread contamination to ecosystems. The present study demonstrated an analysis on accumulation of the toxic pesticide napropamide in six genotypes of alfalfa (Medicago sativa), along with biological parameters and its residues in soils. Soil was treated with napropamide at 3 mg kg(-1) dry soil and alfalfa plants were cultured for 10 or 30 d, respectively. The maximum value for napropamide accumulation is 0.426 mg kg(-1) in shoots and 2.444 mg kg(-1) in roots. The napropamide-contaminated soil with alfalfa cultivation had much lower napropamide concentrations than the control (soil without alfalfa cultivation). Also, the content of napropamide residue in the rhizosphere was significantly lower than that in the non-rhizosphere soil. M. sativa exposed to 3 mg kg(-1) napropamide showed inhibited growth. Further analysis revealed that plants treated with napropamide accumulated more reactive oxygen species (O(2)(-) and H(2)O(2)) and less amounts of chlorophyll. However, not all cultivars showed oxidative injury, suggesting that the alfalfa cultivars display different tolerance to napropamide.