Merck
  • A putative Arabidopsis thaliana glycosyltransferase, At4g01220, which is closely related to three plant cell wall-specific xylosyltransferases, is differentially expressed spatially and temporally.
A putative Arabidopsis thaliana glycosyltransferase, At4g01220, which is closely related to three plant cell wall-specific xylosyltransferases, is differentially expressed spatially and temporally.
Plant science : an international journal of experimental plant biology (2011-03-23)
Jonatan U Fangel, Bent L Petersen, Niels B Jensen, William G T Willats, Antony Bacic, Jack Egelund
ABSTRACT
Plant cell wall polysaccharides are amongst the most complex, heterogeneous and abundant bio-molecules on earth. This makes the biosynthetic enzymes, namely the glycosyltransferases and polysaccharide synthases, important research targets in plant science and biotechnology. As an initial step to characterize At4g01220, a putative Arabidopsis thaliana encoding glycosyltransferases in CAZy GT-family-77 that is similar to three known xylosyltransferases involved in the biosynthesis of the pectic polysaccharide, rhamnogalacturonan II, we conducted an expression analysis. In transgenic Arabidopsis thaliana plants containing a fusion between the At4g01220 promoter and the gusA reporter gene we found the expression to be spatially and developmentally regulated. Analysis of Nicotiana benthamiana transfected with the At2g01220::YFP fusion protein revealed that the fusion protein resided in a Brefeldin A-sensitive compartment consistent with a sub-cellular location in the Golgi apparatus. In addition, in silico expression analysis from the Genevestigator database revealed that At4g01220 was up-regulated upon treatment with isoxaben, an inhibitor of cellulose synthesis, which, together with a co-expression analysis that identified a number of plant cell wall co-related biosynthetic genes, suggests involvement in cell wall biosynthesis with pectin being a prime candidate. The data presented provide insights into the expression, sub-cellular location and regulation of At4g01220 under various conditions and may help elucidate its specific function.