Merck
  • Preparation of molecularly imprinted polymers for the selective recognition of the bioactive polyphenol, (E)-resveratrol.
Preparation of molecularly imprinted polymers for the selective recognition of the bioactive polyphenol, (E)-resveratrol.
Journal of chromatography. A (2011-03-18)
Lachlan J Schwarz, Basil Danylec, Simon J Harris, Reinhard I Boysen, Milton T W Hearn
ABSTRACT
(E)-Resveratrol imprinted polymers have been rationally designed with the aid of molecular modelling and NMR spectroscopic titration techniques to determine the optimal ratio of the template to functional monomer for polymer formation. Based on this approach, (E)-resveratrol imprinted polymers were prepared via non-covalent self-assembly with the functional monomer 4-vinylpyridine (4VP) in a 1:3 molar ratio. Polymerisation in the presence of a cross-linker resulted in rigid block copolymers that had selective capacities towards (E)-resveratrol (e.g. 14 μmol/g) when compared to the non-imprinted reference polymer. The selectivity of these MIPs was also examined using several structurally related polyphenolic compounds to determine the influence of polyphenolic hydroxyl number and position on binding and molecular recognition.
Sigma-Aldrich
4-Vinylpyridine, contains 100 ppm hydroquinone as inhibitor, 95%
Pricing and availability is not currently available.